Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2,5,6-trimethoxybenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

77386-1G 77386-25G 77386-5G 77386-500MG
76918-014EA 91.24 USD InStock
76918-014 76918-016 76918-018 77647-976
3-Bromo-2,5,6-trimethoxybenzoic acid ≥97% research grade
3-Bromo-2,5,6-trimethoxybenzoic acid


Formula: C₁₀H₁₁BrO₅
Storage Temperature: Ambient
MDL Number: MFCD00191620
CAS Number: 101460-22-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty