Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1-(2,2-diethoxyethoxy)-2-fluorobenzene ≥97%

Supplier: AOB CHEM USA
Warning

86051-1G 86051-25G 86051-500MG 86051-5G
76918-088EA 211.12 USD InStock
76918-088 76918-090 76918-092 76918-094
4-Bromo-1-(2,2-diethoxyethoxy)-2-fluorobenzene ≥97%
4-Bromo-1-(2,2-diethoxyethoxy)-2-fluorobenzene
Precaución: for Research Use Only


Formula: C₁₂H₁₆BrFO₃
MW: 307.156 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19168379
CAS Number: 1271673-78-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty