Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichloro-4-methoxy-2-methylbenzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

88452-1G 88452-500MG 88452-5G 88452-25G
76918-154EA 999.37 USD InStock
76918-154 76918-156 76918-158 77660-486
3,5-Dichloro-4-methoxy-2-methylbenzaldehyde ≥97% research grade
3,5-Dichloro-4-methoxy-2-methylbenzaldehyde
Precaución: for Research Use Only


Formula: C₉H₈Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34771522
CAS Number: 67365-50-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty