Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-isopropylbenzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

11153-1G 11153-25G 11153-500MG 11153-5G
76894-516EA 144.29 USD InStock
76894-516 76894-518 76894-520 76894-522
2-Bromo-4-isopropylbenzaldehyde ≥97% research grade
2-Bromo-4-isopropylbenzaldehyde


Formula: C₁₀H₁₁BrO
Storage Temperature: Refrigerator
MDL Number: MFCD18447584
CAS Number: 861897-63-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty