Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzyloxy-5-formylphenylboronic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

12769-1G 12769-25G 12769-5G
76894-902EA 62.6 USD InStock
76894-902 76894-904 76894-906
2-Benzyloxy-5-formylphenylboronic acid ≥95% research grade
2-Benzyloxy-5-formylphenylboronic acid


Formula: C₁₄H₁₃BO₄
MW: 256.062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566593
CAS Number: 1310384-22-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty