Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4,6-dimethoxybenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

128712-1G 128712-500MG 128712-5G
76894-940EA 1148.96 USD InStock
76894-940 76894-942 76894-944
2-Fluoro-4,6-dimethoxybenzoic acid ≥97% research grade
2-Fluoro-4,6-dimethoxybenzoic acid


Formula: C₉H₉FO₄
Storage Temperature: Ambient
CAS Number: 286434-45-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty