Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-3-(methoxymethoxy)-4-(methylthio)pyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

129219-1G 129219-25G 129219-500MG 129219-5G
76894-954EA 151.71 USD InStock
76894-954 76894-956 76894-958 76894-960
2,6-Dichloro-3-(methoxymethoxy)-4-(methylthio)pyridine ≥97% research grade
2,6-Dichloro-3-(methoxymethoxy)-4-(methylthio)pyridine


Formula: C₈H₉Cl₂NO₂S
Storage Temperature: Refrigerator
CAS Number: 2918958-46-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty