Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-(methylsulfonyl)benzeneboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

13621-1G 13621-25G 13621-500MG 13621-5G
76895-104EA 224.92 USD InStock
76895-104 76895-106 76895-108 76895-110
2-Fluoro-4-(methylsulfonyl)benzeneboronic acid ≥97% research grade
2-Fluoro-4-(methylsulfonyl)benzeneboronic acid


Formula: C₇H₈BFO₄S
MW: 218.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027248
CAS Number: 957060-85-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty