Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(3,5-Dichlorophenylcarbamoyl)-4-fluorophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

14088-1G 14088-25G 14088-5G
76895-232EA 64.72 USD InStock
76895-232 76895-234 76895-236
3-(3,5-Dichlorophenylcarbamoyl)-4-fluorophenylboronic acid ≥97%
3-(3,5-Dichlorophenylcarbamoyl)-4-fluorophenylboronic acid


Formula: C₁₃H₉BCl₂FNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11856004
CAS Number: 1451393-28-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty