Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Bis(trifluoromethyl)benzeneboronic acid ≥95%

Supplier: AOB CHEM USA
Warning

14630-1G 14630-25G 14630-5G
76895-584EA 62.6 USD InStock
76895-584 76895-586 76895-588
2,5-Bis(trifluoromethyl)benzeneboronic acid ≥95%
2,5-Bis(trifluoromethyl)benzeneboronic acid
Precaución: for Research Use Only


Formula: C₈H₅BF₆O₂
MW: 257.926 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038223
CAS Number: 196083-18-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty