Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dibromo-4-methylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

15182-1G 15182-25G 15182-500MG 15182-5G
76895-734EA 153.84 USD InStock
76895-734 76895-736 76895-738 76895-740
2,6-Dibromo-4-methylbenzoic acid ≥97%
2,6-Dibromo-4-methylbenzoic acid


Formula: C₈H₆Br₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11847286
CAS Number: 128733-74-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty