Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenoxy-5-(trifluoromethyl)phenylamine ≥97% research grade

Supplier: AOB CHEM USA
Warning

154911-1G 154911-25G 154911-500MG 154911-5G
76896-000EA 185.66 USD InStock
76896-000 76896-002 76896-004 76896-006
2-Phenoxy-5-(trifluoromethyl)phenylamine ≥97% research grade
2-Phenoxy-5-(trifluoromethyl)phenylamine


Formula: C₁₃H₁₀F₃NO
Storage Temperature: Ambient
MDL Number: MFCD08688474
CAS Number: 50594-29-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty