Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-(trifluoromethoxy)phenyboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

15743-1G 15743-25G 15743-5G
76896-142EA 62.6 USD InStock
76896-142 76896-144 76896-146
2-Bromo-6-(trifluoromethoxy)phenyboronic acid ≥97% research grade
2-Bromo-6-(trifluoromethoxy)phenyboronic acid


Formula: C₇H₅BBrF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD11111908
CAS Number: 1451393-18-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty