Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Carboxy-5-chlorophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

16034-1G 16034-25G 16034-500MG 16034-5G
76896-306EA 213.25 USD InStock
76896-306 76896-308 76896-310 76896-312
2-Carboxy-5-chlorophenylboronic acid ≥97%
2-Carboxy-5-chlorophenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₆BClO₄
MW: 200.384 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08235091
CAS Number: 874290-67-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty