Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-3-(trifluoromethyl)phenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

16052-1G 16052-25G 16052-5G 16052-500MG
76896-322EA 96.55 USD InStock
76896-322 76896-324 76896-326 77644-716
2,6-Dichloro-3-(trifluoromethyl)phenylboronic acid ≥97% research grade
2,6-Dichloro-3-(trifluoromethyl)phenylboronic acid


Formula: C₇H₄BCl₂F₃O₂
MW: 258.818 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526369
CAS Number: 1027059-21-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty