Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-4-METHYL-3-PYRIDINYLBORONIC ACID ≥97%

Supplier: AOB CHEM USA
Warning

16707-1G 16707-25G 16707-500MG 16707-5G
76896-528EA 409.51 USD InStock
76896-528 76896-530 76896-532 76896-534
2-Methoxy-4-METHYL-3-PYRIDINYLBORONIC ACID ≥97%
2-Methoxy-4-METHYL-3-PYRIDINYLBORONIC ACID


Formula: C₇H₁₀BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD07374996
CAS Number: 1029654-21-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty