Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-fluoro-3-hydroxyphenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

17079-1G 17079-25G 17079-500MG 17079-5G
76896-698EA 165.51 USD InStock
76896-698 76896-700 76896-702 76896-704
2-Bromo-6-fluoro-3-hydroxyphenylboronic acid ≥97% research grade
2-Bromo-6-fluoro-3-hydroxyphenylboronic acid


Formula: C₆H₅BBrFO₃
MW: 234.816 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16295246
CAS Number: 1451392-83-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty