Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-iodo-3-methylpyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

17571-1G 17571-25G 17571-500MG 17571-5G
76896-890EA 144.29 USD InStock
76896-890 76896-892 76896-894 76896-896
2-Fluoro-5-iodo-3-methylpyridine ≥97% research grade
2-Fluoro-5-iodo-3-methylpyridine


Formula: C₆H₅FIN
MW: 237.013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839280
CAS Number: 205245-17-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty