Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-fluorophenacyl bromide ≥97%

Supplier: AOB CHEM USA
Warning

17694-1G 17694-25G 17694-500MG 17694-5G
76896-946EA 289.63 USD InStock
76896-946 76896-948 76896-950 76896-952
4-Bromo-3-fluorophenacyl bromide ≥97%
4-Bromo-3-fluorophenacyl bromide
Precaución: for Research Use Only


Formula: C₈H₅Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD11847180
CAS Number: 1003879-02-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty