Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-methoxynicotinic acid ≥97%

Supplier: AOB CHEM USA
Warning

18763-1G 18763-25G 18763-5G
76897-320EA 96.55 USD InStock
76897-320 76897-322 76897-324
2-Chloro-6-methoxynicotinic acid ≥97%
2-Chloro-6-methoxynicotinic acid
Precaución: for Research Use Only


Formula: C₇H₆NO₃Cl
Storage Temperature: Refrigerator
MDL Number: MFCD08236845
CAS Number: 503000-87-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty