Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-4-methylphenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

19171-1G 19171-25G 19171-500MG 19171-5G
76897-502EA 201.58 USD InStock
76897-502 76897-504 76897-506 76897-508
2-Methoxy-4-methylphenylboronic acid ≥97% research grade
2-Methoxy-4-methylphenylboronic acid


Formula: C₈H₁₁BO₃
MW: 165.982 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18072608
CAS Number: 198211-79-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty