Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Carboxythiophene-3-boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

19914-1G 19914-25G 19914-5G
76897-706EA 88.06 USD InStock
76897-706 76897-708 76897-710
2-Carboxythiophene-3-boronic acid ≥97% research grade
2-Carboxythiophene-3-boronic acid


Formula: C₅H₅BO₄S
MW: 171.967 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06203318
CAS Number: 5503-74-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty