Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-5-(methoxycarbonyl)phenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

20691-1G 20691-25G 20691-500MG 20691-5G
76898-128EA 168.69 USD InStock
76898-128 76898-130 76898-132 76898-134
2-Methoxy-5-(methoxycarbonyl)phenylboronic acid ≥97%
2-Methoxy-5-(methoxycarbonyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₁BO₅
MW: 209.9916 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09800877
CAS Number: 221006-63-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty