Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3,5-dimethylbenzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

21018-1G 21018-25G 21018-500MG 21018-5G
76898-274EA 162.32 USD InStock
76898-274 76898-276 76898-278 76898-280
2-Fluoro-3,5-dimethylbenzaldehyde ≥97% research grade
2-Fluoro-3,5-dimethylbenzaldehyde


Formula: C₉H₉FO
Storage Temperature: Ambient
MDL Number: MFCD28550880
CAS Number: 1780780-18-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty