Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Difluoro-4-formylphenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

21119-1G 21119-25G 21119-500MG 21119-5G
76898-294EA 210.06 USD InStock
76898-294 76898-296 76898-298 76898-300
2,5-Difluoro-4-formylphenylboronic acid ≥97% research grade
2,5-Difluoro-4-formylphenylboronic acid


Formula: C₇H₅BF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD18383627
CAS Number: 1228828-19-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty