Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Formyl-5-isopropylphenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

21228-1G 21228-25G 21228-500MG 21228-5G
76898-322EA 215.37 USD InStock
76898-322 76898-324 76898-326 76898-328
2-Formyl-5-isopropylphenylboronic acid ≥97% research grade
2-Formyl-5-isopropylphenylboronic acid


Formula: C₁₀H₁₃BO₃
MW: 192.019 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19105372
CAS Number: 1451390-89-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty