Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloro-N-methoxy-N-methylisonicotinamide ≥97% research grade

Supplier: AOB CHEM USA
Warning

21822-1G 21822-25G 21822-500MG 21822-5G
76898-494EA 213.25 USD InStock
76898-494 76898-496 76898-498 76898-500
2,5-Dichloro-N-methoxy-N-methylisonicotinamide ≥97% research grade
2,5-Dichloro-N-methoxy-N-methylisonicotinamide


Formula: C₈H₈Cl₂N₂O₂
MW: 235.067 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15526781
CAS Number: 864674-17-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty