Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Fluoro-3-methylpyridin-4-yl)boronic acid ≥97%

Supplier: AOB CHEM USA
Warning

21991-1G 21991-25G 21991-500MG 21991-5G
76898-548EA 215.37 USD InStock
76898-548 76898-550 76898-552 76898-554
(2-Fluoro-3-methylpyridin-4-yl)boronic acid ≥97%
(2-Fluoro-3-methylpyridin-4-yl)boronic acid
Precaución: for Research Use Only


Formula: C₆H₇BFNO₂
MW: 154.935 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611287
CAS Number: 1451391-34-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty