Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(trifluoromethoxy)benzoic acid methyl ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

22141-1G 22141-25G 22141-500MG 22141-5G
76898-618EA 195.21 USD InStock
76898-618 76898-620 76898-622 76898-624
2-Bromo-4-(trifluoromethoxy)benzoic acid methyl ester ≥97% research grade
2-Bromo-4-(trifluoromethoxy)benzoic acid methyl ester


Formula: C₉H₆BrF₃O₃
MW: 299.041 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698162
CAS Number: 1214366-07-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty