Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-(sec-butoxy)naphthalene ≥95%

Supplier: AOB CHEM USA
Warning

22498-1G 22498-25G 22498-5G
76898-720EA 64.72 USD InStock
76898-720 76898-722 76898-724
2-Bromo-6-(sec-butoxy)naphthalene ≥95%
2-Bromo-6-(sec-butoxy)naphthalene


Formula: C₁₄H₁₅BrO
Storage Temperature: Ambient
MDL Number: MFCD18360988
CAS Number: 1492479-97-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty