Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-methoxy-4-methylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

22803-1G 22803-25G 22803-500MG 22803-5G
76898-904EA 290.69 USD InStock
76898-904 76898-906 76898-908 76898-910
2-Fluoro-6-methoxy-4-methylphenylboronic acid ≥97%
2-Fluoro-6-methoxy-4-methylphenylboronic acid


Formula: C₈H₁₀BFO₃
Storage Temperature: Ambient
MDL Number: MFCD22543654
CAS Number: 1451392-08-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty