Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-methoxybenzyl alcohol ≥95%

Supplier: AOB CHEM USA
Warning

23422-1G 23422-25G 23422-500MG 23422-5G
76899-048EA 185.66 USD InStock
76899-048 76899-050 76899-052 76899-054
2-Chloro-5-methoxybenzyl alcohol ≥95%
2-Chloro-5-methoxybenzyl alcohol


Formula: C₈H₉ClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD12024393
CAS Number: 101252-66-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty