Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3,4-difluorophenol ≥95% research grade

Supplier: AOB CHEM USA
Warning

23534-1G 23534-25G 23534-500MG 23534-5G
76899-104EA 260.99 USD InStock
76899-104 76899-106 76899-108 76899-110
2-Chloro-3,4-difluorophenol ≥95% research grade
2-Chloro-3,4-difluorophenol


Formula: C₆H₃ClF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD11501038
CAS Number: 1159512-40-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty