Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2,4-Difluorophenyl)benzaldehyde ≥95% research grade

Supplier: AOB CHEM USA
Warning

23927-1G 23927-25G 23927-500MG 23927-5G
76899-250EA 201.58 USD InStock
76899-250 76899-252 76899-254 76899-256
3-(2,4-Difluorophenyl)benzaldehyde ≥95% research grade
3-(2,4-Difluorophenyl)benzaldehyde


Formula: C₁₃H₈F₂O
Storage Temperature: Refrigerator
MDL Number: MFCD04117389
CAS Number: 728918-77-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty