Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-isopropoxypyridine ≥97%

Supplier: AOB CHEM USA
Warning

24416-1G 24416-25G 24416-500MG 24416-5G
76899-452EA 213.25 USD InStock
76899-452 76899-454 76899-456 76899-458
2-Chloro-6-isopropoxypyridine ≥97%
2-Chloro-6-isopropoxypyridine
Precaución: for Research Use Only


Formula: C₈H₁₀ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD11113405
CAS Number: 89481-98-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty