Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-6-(methylthio)-3-pyridylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

24586-1G 24586-25G 24586-500MG 24586-5G
76899-562EA 260.99 USD InStock
76899-562 76899-564 76899-566 76899-568
2-Methyl-6-(methylthio)-3-pyridylboronic acid ≥97% research grade
2-Methyl-6-(methylthio)-3-pyridylboronic acid


Formula: C₇H₁₀BNO₂S
MW: 183.036 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22543735
CAS Number: 1451392-60-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty