Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-5-phenylpyridine ≥97%

Supplier: AOB CHEM USA
Warning

24890-1G 24890-25G 24890-500MG 24890-5G
76899-724EA 231.28 USD InStock
76899-724 76899-726 76899-728 76899-730
2-Methoxy-5-phenylpyridine ≥97%
2-Methoxy-5-phenylpyridine
Precaución: for Research Use Only


Formula: C₁₂H₁₁NO
MW: 185.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044373
CAS Number: 53698-47-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty