Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-5-(methoxymethyl)phenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

25250-1G 25250-500MG 25250-5G 25250-25G
76899-920EA 485.9 USD InStock
76899-920 76899-922 76899-924 77658-230
2-Methoxy-5-(methoxymethyl)phenylboronic acid ≥97% research grade
2-Methoxy-5-(methoxymethyl)phenylboronic acid


Formula: C₉H₁₃BO₄
MW: 196.008 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14687258
CAS Number: 1332504-32-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty