Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-chlorophenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

25618-1G 25618-25G 25618-500MG 25618-5G
76900-032EA 165.51 USD InStock
76900-032 76900-034 76900-036 76900-038
2-Amino-5-chlorophenylboronic acid pinacol ester ≥97%
2-Amino-5-chlorophenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₂H₁₇BClNO₂
Storage Temperature: Ambient
MDL Number: MFCD06795672
CAS Number: 1073371-77-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty