Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-fluoro-3-iodopyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

25752-1G 25752-25G 25752-500MG 25752-5G
76900-092EA 213.25 USD InStock
76900-092 76900-094 76900-096 76900-098
2-Chloro-4-fluoro-3-iodopyridine ≥97% research grade
2-Chloro-4-fluoro-3-iodopyridine


Formula: C₅H₂ClFIN
Storage Temperature: Refrigerator
MDL Number: MFCD19686167
CAS Number: 1271477-28-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty