Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-iodo-3-methoxypyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

25902-1G 25902-25G 25902-500MG 25902-5G
76900-162EA 171.87 USD InStock
76900-162 76900-164 76900-166 76900-168
2-Fluoro-4-iodo-3-methoxypyridine ≥97% research grade
2-Fluoro-4-iodo-3-methoxypyridine


Formula: C₆H₅FINO
MW: 253.013 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16611136
CAS Number: 1227580-53-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty