Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(trifluoromethoxy)phenylboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

26179-1G 26179-25G 26179-500MG 26179-5G
76900-276EA 224.92 USD InStock
76900-276 76900-278 76900-280 76900-282
2-Bromo-4-(trifluoromethoxy)phenylboronic acid pinacol ester ≥97% research grade
2-Bromo-4-(trifluoromethoxy)phenylboronic acid pinacol ester


Formula: C₁₃H₁₅BBrF₃O₃
MW: 366.967 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29915586
CAS Number: 2121514-01-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty