Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Dimethylamino-4-bromothiazole ≥97% research grade

Supplier: AOB CHEM USA
Warning

26923-1G 26923-25G 26923-500MG 26923-5G
76900-540EA 170.81 USD InStock
76900-540 76900-542 76900-544 76900-546
2-Dimethylamino-4-bromothiazole ≥97% research grade
2-Dimethylamino-4-bromothiazole


Formula: C₅H₇BrN₂S
MW: 207.091 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878986
CAS Number: 209260-76-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty