Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-5-phenylbenzaldehyde ≥95% research grade

Supplier: AOB CHEM USA
Warning

27045-1G 27045-25G 27045-500MG 27045-5G
76900-570EA 370.26 USD InStock
76900-570 76900-572 76900-574 76900-576
2-Methyl-5-phenylbenzaldehyde ≥95% research grade
2-Methyl-5-phenylbenzaldehyde


Formula: C₁₄H₁₂O
MW: 196.244 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24674947
CAS Number: 2928-45-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty