Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylbiphenyl-4-ylboronic acid ≥95%

Supplier: AOB CHEM USA
Warning

27199-1G 27199-25G 27199-500MG 27199-5G
76900-624EA 389.36 USD InStock
76900-624 76900-626 76900-628 76900-630
2-Methylbiphenyl-4-ylboronic acid ≥95%
2-Methylbiphenyl-4-ylboronic acid


Formula: C₁₃H₁₃BO₂
Storage Temperature: Refrigerator
MDL Number: MFCD28009348
CAS Number: 1383628-42-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty