Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-methoxybenzyl alcohol ≥97%

Supplier: AOB CHEM USA
Warning

27458-1G 27458-25G 27458-5G
76900-696EA 96.55 USD InStock
76900-696 76900-698 76900-700
2-Chloro-4-methoxybenzyl alcohol ≥97%
2-Chloro-4-methoxybenzyl alcohol


Formula: C₈H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD16037274
CAS Number: 334018-24-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty