Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3,5-dimethylbenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

27580-1G 27580-25G 27580-500MG 27580-5G
76900-720EA 350.1 USD InStock
76900-720 76900-722 76900-724 76900-726
2-Chloro-3,5-dimethylbenzoic acid ≥97% research grade
2-Chloro-3,5-dimethylbenzoic acid


Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11846006
CAS Number: 90649-75-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty