Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-(trifluoromethyl)isonicotinaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

30032-1G 30032-25G 30032-500MG 30032-5G
76901-422EA 439.22 USD InStock
76901-422 76901-424 76901-426 76901-428
2-Chloro-6-(trifluoromethyl)isonicotinaldehyde ≥97% research grade
2-Chloro-6-(trifluoromethyl)isonicotinaldehyde


Formula: C₇H₃ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD13188805
CAS Number: 1060807-47-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty