Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-4-trifluromethyl methylbenzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

30137-1G 30137-25G 30137-500MG 30137-5G
76901-436EA 173.99 USD InStock
76901-436 76901-438 76901-440 76901-442
2-Methoxy-4-trifluromethyl methylbenzoate ≥97% research grade
2-Methoxy-4-trifluromethyl methylbenzoate


Formula: C₁₀H₉F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD7781286
CAS Number: 286441-66-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty