Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethoxypyridine-4-carboxaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

30399-1G 30399-500MG 30399-5G 30399-25G
76901-500EA 485.9 USD InStock
76901-500 76901-502 76901-504 77654-846
2-Ethoxypyridine-4-carboxaldehyde ≥97%
2-Ethoxypyridine-4-carboxaldehyde


Formula: C₈H₉NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD17012139
CAS Number: 1211587-25-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty